missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ Comfort Pad for S-950 Premium Head Suspension

For use with S-950 Premium Head Suspension

$35.23 - $71.00

Specifications

Color Gray
Disposable Reusable
Product Type Head Suspension
For Use With (Application) Cleaning, Electrical, Facility Maintenance, Welding
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-805-616
View Documents
3M™
7000127438
Standard
Pack of 5 Each for $35.23
Case of 4 Pack for $140.92
Only null left
 
19-805-617
View Documents
3M™
7100010371
Small
Pack of 5 Each for $71.00
Case of 4 Pack for $284.00
Only null left
 
Description

Description

  • For use with S-950 Premium Head Suspension
  • Wrapped around headband to create more comfortable fit and to absorb sweat
  • 3M Catalog# S-953 , 3M Catalog# S-954

Specifications

Specifications

Gray
Reusable
Head Suspension
Cleaning, Electrical, Facility Maintenance, Welding
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'