missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ Comfort Pad for S-950 Premium Head Suspension

Catalog No. 19805617
Change view
Click to view available options
Size:
Standard
Small
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19805617 Small
19805616 Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19805617 Supplier 3M™ Supplier No. 7100010371

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with S-950 Premium Head Suspension

  • For use with S-950 Premium Head Suspension
  • Wrapped around headband to create more comfortable fit and to absorb sweat
  • 3M Catalog# S-953 , 3M Catalog# S-954

Specifications

Color Gray
Disposable Reusable
Product Type Head Suspension
Size Small
For Use With (Application) Cleaning, Electrical, Facility Maintenance, Welding
For Use With (Equipment) Premium Head Suspension
Model S-954