Learn More
Propper Verline Indicator Tape
Propper Verline Indicator Tape secures packages during steam sterilization and confirms exposure to the sterilant after processing. Beige Indicator Tape features distinct green to black color change after exposure to steam.
$1165.14 - $1178.32
Specifications
Autoclavable Autoclavable
Description Indicator tape printed with green stripes
Adhesive Type Synthetic
Coating Synthetic Resin
Color Tan
View More Specs
Products 2
Catalog Number Mfr. No. Width (English) Price Quantity  
Catalog Number Mfr. No. Width (English) Price Quantity  
15375172
View Documents
Propper
26810600
1 in.
Case of 36 Each for $1,178.32
Only null left
15375173
View Documents
Propper
26810500
0.75 in.
Case of 48 Each for $1,165.14
Only null left
Description
Description
  • Propper Verline™ Beige Autoclave Indicator Tape is a pressure-sensitive sterilization tape used to distinguish between processed and unprocessed items.
  • Verline™ is a beige autoclave indicator tape used in steam sterilizers. Featuring lead-free indicator ink, placed in green stripes that turn black after exposure to a steam sterilization cycle.
  • Verline™ tape's strong adhesive and even coating allow for even dispensing and easy application. Specifically formulated universal adhesive provides secure bonding with fabric and non-woven wraps, paper, metal, and glass, yet provides easy removal without traces of residue.
  • AAMI/ISO 11140-1 Type 1 Compliant Process Indicator Tape
  • Not Made with Natural Rubber Latex
Specifications
Specifications
Autoclavable
Synthetic
Tan
Crepe Paper
Non-sterile
121°C for 30 min., 132°C for 4 min., 135°C for 3 min.
55 m
Indicator tape printed with green stripes
Synthetic Resin
Roll
Tape
250°F for 30 min., 270°F for 4 min., 275°F for 3 min.
60 yd.