missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Veriti™ Thermal Cycler, 96-well Fast

Catalog No. A27924
Encompass
Change view
Click to view available options
Warranty:
5 years of total service coverage with instrument rapid exchange
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Warranty
A27924 5 years of total service coverage with instrument rapid exchange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A27924 Supplier Applied Biosystems™ Supplier No. A27924

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Applied Biosystems Veriti Thermal Cycler delivers the proven reliability of Applied Biosystems instruments. With the added control of VeriFlex technology, you have six independent temperature blocks that provide precise control over your PCR optimization. The Veriti 96-Well Fast Thermal Cycler is equipped with a Fast 0.1-mL block configuration.

The Applied Biosystems Veriti Thermal Cycler delivers the proven reliability of Applied Biosystems instruments. With the added control of VeriFlex technology, you have six independent temperature blocks that provide precise control over your PCR optimization. Additionally, the color touch screen simplifies setup and use of the Veriti Thermal Cycler. Optional setups for fast or standard PCR methods provide with flexibility to shorten your PCR cycling times. The Veriti 96-Well Fast Thermal Cycler is equipped with a Fast 0.1-mL block configuration.

Features include:

  • Innovative VeriFlex Blocks allow for precise PCR optimization and enable you to run up to six independent assays
  • Fast 0.1-mL block format is compatible with many downstream applications
  • Standard and fast enabled cycling to address your current and future PCR needs
  • Easy-to-use graphical interface (6.5 inch VGA Touch Screen)
  • Convenient protocol transfer from one Veriti Thermal Cycler to another using USB
  • Reduced PCR cycling time when using faster ramp rates
  • Optional VeritiLink Remote Management Software provides you with peace of mind, allowing you to remotely manage more than 50 cyclers from your PC or internet-compatible phone

VeriFlex Blocks: Enhanced PCR Functionality

The Veriti Thermal Cycler features innovative VeriFlex Blocks for enhanced PCR functionality. Six separate peltier blocks provide maximal versatility and flexibility and offer two key benefits: 1) PCR Optimization: each block can be set with a specific temperature which is ideal for precise control over PCR optimization 2) Run more experiments: the six peltier blocks can also be used to run up to six different annealing temperatures in the same run. With this enhanced functionality, the Veriti Thermal Cycler with VeriFlex blocks is the ideal thermal cycler for any lab.

Control at your fingertips

The powerful, yet simple to operate user interface on the Veriti system is driven by the 6.5 inch (16.51 cm) VGA color touch screen. The large screen allows for easy viewing of your temperature profiles. Additionally, the large navigation buttons put programming of the Veriti Thermal Cycler at your finger tips. Setup and navigation of the Veriti Thermal Cycler does not require the use of a stylus or mouse.

Fast setup, fast results

The Veriti system has two different options for method navigation. For quick setup, you may select one of the pre-programmed methods to run. If you prefer to program your own methods, simply touch the step you would like to setup and then use the keypad to enter thermal cycling steps. After completing a setup, you have the choice of saving the method to the instrument or to a USB memory stick.

Control your time: Go fast when you want

With the Veriti Thermal Cycler you get two thermal cyclers in one, a standard thermal cycler with its better than gradient feature, and a fast enabled thermal cycler. So whether you have a current need to run fast PCR and shorten your PCR cycling times, or a possible future application need for fast PCR, the Veriti Thermal Cycler provides you with the flexibility to go fast when you want.

Applied Biosystems PCR plastics

Applied Biosystems PCR plastics have been designed and validated to work with our thermal cyclers for more than 25 years. Options for every format and throughput need are available to ensure consistent, precise results for any experiment. To find the best match please visit our Plastics Selection Guide.

Specifications

Format 0.1 mL Tubes, 96-well Plate
Thermal Accuracy ±0.25°C (35°C to 99.9°C)
Sample Ramp Rate ±4.25°C/sec.
Temperature Range (Metric) 4°C to 99.9°C
Reaction Volume Range 10 to 30 μL
Content And Storage Veriti 96-well Fast Thermal Cycler + 3 year REX extended warranty
High-throughput Compatibility High-throughput Compatible
For Use With (Equipment) Veriti™ Thermal Cycler
Thermal Uniformity <0.5°C (20 sec. after reaching 95°C)
Reaction Speed Fast, Standard
Dimensions 19.1 x 9.3 x 9.6 in. (48.5 x 23.7 x 24.5 cm)
Capacity 96-well, 0.1 mL Tubes
Voltage 100/240 V
Product Line Veriti
Peak Block Ramp Rate 5.0°C/sec.
Display Type 6.5 in. VGA 32K Color Touchscreen
Product Type Thermal Cycler
Warranty 5 years of total service coverage with instrument rapid exchange
Thermal Range 0°C to 100°C
For Use With (Application) PCR
Programs Auto re-start (after Power Outages), Program Overwrite Protection
User Interface USB
Max. Ramp Rate 5°C/sec. (Block), 4.25°C/sec. (Sample)
Quantity System + 3 yr REX ext warranty
Memory USB and On-board
Remarks VeriFlex™ Blocks: 25°C (5°C Zone-to-Zone), Fast 0.1 mL format and sample block enabled to run fast chemistry
Tm Calculator Menu Driven through Touchscreen
Warranty Services Not Included User-replaceable Parts, Planned Maintenance
Warranty Response Time Replacement instrument shipped in 1 business day and received within 3 days
Weight 25 lb. (11.4 kg)
Block Configurations 96-well, 6-Zone VeriFlex Block
Show More Show Less
Have you tested the Veriti and VeritiPro Thermal Cyclers in low humidity conditions (below 20%)?

The recommended conditions are determined to ensure no static discharge (low humidity) occurs.
Recommended Conditions:
- Veriti: 20-80% humidity
- VeritiPro: 15-80% humidity
We have not tested Veriti and VeritiPro thermal cyclers in low humidity, below 20%, therefore we cannot recommend using them in those conditions.

What is the optimal ramp rate for my thermal cycler?

There is no universal ramp rate for a thermal cycler because both the block and sample ramp rates are different for each thermal cycler. The sample ramp rate changes with the volume of the sample since more time and energy are required to heat higher sample volumes. The ramp rate may also need to be changed for a particular PCR assay or primer pair if you are trying to use the same PCR assay on two different thermal cyclers, for example.

As a reference, the block ramp rate commonly used with DreamTaq Polymerase on the MiniAmp thermal cycler is 3.0 C/sec. In a 50 µL PCR reaction, this translates to a sample ramp rate of ~1 C/sec.

Please refer to the following application note for further information about thermocycler ramp rates: https://assets.thermofisher.com/TFS-Assets/LSG/brochures/Thermal_Cycler_Ramp_RatesAppNote.pdf

For Research Use Only. Not for use in diagnostic procedures.