missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Vanadium Standard for ICP 10 g/L in 5% HNO3, Ricca Chemical

Catalog No. RV010496100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010496100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RV010496100 Supplier Ricca Chemical Company Supplier No. RV010496100N

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 10 g/L in 5% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Specifications

Packaging Natural LDPE Bottle
CAS 7803-55-6, 7697-37-2, 7732-18-5
CAS Min % 90.85%, 4.90%, 2.25%
CAS Max % 94.55%, 5.10%, 2.35%
Specific Gravity 1
Physical Form Liquid
Molecular Formula H4NO3V
MDL Number MFCD00011430
UN Number UN3289
Color Colorless
Melting Point Approximately 0°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Standard
InChI Key YBVKNHXQSRDWAA-UHFFFAOYSA-M
SMILES N.O[V](=O)=O
IUPAC Name vanadiumoylol amine
Molecular Weight (g/mol) 116.98
Quantity 100 mL
PubChem CID 516859
Grade High Purity
Chemical Name or Material Vanadium ICP Standard
Show More Show Less