missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Tungsten Standard for ICP 10 g/L in 10% NH3, Ricca Chemical

Catalog No. RV010497100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010497100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RV010497100 Supplier Ricca Chemical Company Supplier No. RV010497100N

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 10 g/L in 10% NH3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Specifications

Packaging Natural LDPE Bottle
CAS 11140-77-5, 1336-21-6, 7732-18-5
CAS Min % 86.84%, 9.80%, 1.36%
CAS Max % 90.38%, 10.20%, 1.42%
Specific Gravity 1
Physical Form Liquid
Molecular Formula H40N10O41W12
UN Number UN2672
Color Colorless
pH <2
Melting Point Approximately 0°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Standard
InChI Key LMAJGSPZJYEENT-UHFFFAOYSA-D
SMILES N.N.N.N.N.N.N.N.N.N.O[W]12(=O)O[W]3(=O)(O[W](=O)(O3)(O[W]4(=O)(O[W](=O)(O4)(O[W]5(=O)(O[W]6(=O)(O5)O[W]7(=O)(O6)O[W](=O)(O7)(O[W]8(=O)(O[W](=O)(O8)(O[W](=O)(O1)(O2)O)O)O)O)O)O)O)O)O
Molecular Weight (g/mol) 3042.429
Quantity 100 mL
PubChem CID 71306883
Grade High Purity
Chemical Name or Material Tungsten ICP Standard
Show More Show Less