missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Titanium Standard for ICP/MS 10 ppm in 2% HNO3/HF, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Supplier: Ricca Chemical Company RV010629100N
Description
- 10 ppm in 2% HNO3/HF
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
| Titanium ICP-MS Standard | |
| 16962-40-6,7697-37-2,7664-39-3,16962-40-6 | |
| 99.76%,2.04%,0.20%,0.00% | |
| Liquid | |
| Approximately 0°C | |
| Miscible with water | |
| Standard | |
| High Purity |
| Natural LDPE Bottle | |
| 95.84%,1.96%,0.20%,0.00% | |
| 1 | |
| Colorless | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Calibration Standard | |
| 100 mL |