missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Tin Standard for ICP/MS 10 ppm in 1% HNO3/HF, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$435.56 - $435.56
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010623100
|
Ricca Chemical Company
RV010623100N |
100 mL |
Each for $435.56
|
|
|||||
Description
- 10 ppm in 1% HNO3/HF
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 7783-62-2 |
Specifications
| Natural LDPE Bottle | |
| 96.53%,0.98%,0.49%,0.00% | |
| 1 | |
| Approximately 0°C | |
| 100 mL |
| 7783-62-2,7697-37-2,7664-39-3,7783-62-2 | |
| 100%,1.02%,0.51%,0.00% | |
| Liquid | |
| Standard | |
| High Purity |