missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Tin Standard for ICP 1000 ppm in 20% HCl, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

$479.27 - $1084.63

Chemical Identifiers

CAS 7783-62-2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RV010316100
View Documents
Ricca Chemical Company
RV010316100N
100 mL
Each for $479.27
Only null left
 
RV010316250
View Documents
Ricca Chemical Company
RV010316250N
250 mL
Each for $762.07
Only null left
 
RV010316500
View Documents
Ricca Chemical Company
RV010316500N
500 mL
Each for $1,084.63
Only null left
 
Description

Description

  • 1000 ppm in 20% HCl
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Chemical Identifiers

7783-62-2

Specifications

Natural LDPE Bottle
78.18%,19.60%,0.22%
1
UN1789
Standard
High Purity
7783-62-2,7647-01-0,7783-62-2
81.38%,20.40%,0.22%
Liquid
Approximately 0°C
100 mL
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'