missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Tin (Sn) Standard for AAS 1000 ppm in 1% HNO3/1% HF, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

$220.66 - $388.69

Chemical Identifiers

CAS 7783-62-2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RV010052100
View Documents
Ricca Chemical Company
RV010052100N
100 mL N/A
Only null left
 
RV010052500
View Documents
Ricca Chemical Company
RV010052500N
500 mL N/A
Only null left
 
Description

Description

  • 1000 ppm in 1% HNO3/1% HF
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Chemical Identifiers

7783-62-2

Specifications

Natural LDPE Bottle
95.88%,0.98%,0.16%
1.01
UN3287
100 mL
7783-62-2,7697-37-2,7783-62-2
99.80%,1.02%,0.16%
Liquid
Approximately 0°C
High Purity
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'