missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Tellurium Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$438.30 - $1225.97
Chemical Identifiers
| CAS | 7803-68-1 |
|---|---|
| Molecular Formula | H6O6Te |
| Molecular Weight (g/mol) | 229.642 |
| InChI Key | FXADMRZICBQPQY-UHFFFAOYSA-N |
| PubChem CID | 62686 |
| ChEBI | CHEBI:30461 |
| SMILES | O[Te](O)(O)(O)(O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010356100
|
Ricca Chemical Company
RV010356100N |
100 mL |
Each for $438.30
|
|
|||||
|
RV010356250
|
Ricca Chemical Company
RV010356250N |
250 mL |
Each for $854.53
|
|
|||||
|
RV010356500
|
Ricca Chemical Company
RV010356500N |
500 mL |
Each for $1,225.97
|
|
|||||
Description
- 1000 ppm in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 7803-68-1 | |
| 229.642 | |
| 62686 | |
| O[Te](O)(O)(O)(O)O |
| H6O6Te | |
| FXADMRZICBQPQY-UHFFFAOYSA-N | |
| CHEBI:30461 |
Specifications
| Natural LDPE Bottle | |
| 95.89%,1.96%,0.15% | |
| 1 | |
| H6O6Te | |
| Standard | |
| O[Te](O)(O)(O)(O)O | |
| 100 mL | |
| CHEBI:30461 |
| 7803-68-1,7697-37-2,7732-18-5 | |
| 99.81%,2.04%,0.15% | |
| Liquid | |
| Approximately 0°C | |
| FXADMRZICBQPQY-UHFFFAOYSA-N | |
| 229.642 | |
| 62686 | |
| High Purity |