missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Sulfur Standard for ICP/MS 10 ppm in H2O, Ricca Chemical

Catalog No. RV010617100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010617100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RV010617100 Supplier Ricca Chemical Company Supplier No. RV010617100N

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
PubChem CID 6097028
ChEBI CHEBI:62946
IUPAC Name sulfuric acid diamine
SMILES N.N.OS(O)(=O)=O

Specifications

Packaging Natural LDPE Bottle
CAS Min % 98%, 0.00%
CAS Max % 100%, 0.00%
Specific Gravity 1
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Standard
Quantity 100 mL
Grade High Purity
Chemical Name or Material Sulfur ICP-MS Standard
Show More Show Less