missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Strontium Ionization Buffer for AAS 2% Sr in 2% HCl, Ricca Chemical
Description
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
| Packaging | Natural LDPE Bottle |
| CAS | 7732-18-5, 10476-85-4, 7647-01-0 |
| CAS Min % | 92.49%, 3.55%, 1.96% |
| CAS Max % | 96.27%, 3.69%, 2.04% |
| Specific Gravity | 1.02 |
| Physical Form | Liquid |
| Melting Point | Approximately 0°C |
| Quantity | 100 mL |
| Grade | High Purity |