missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Scandium (Sc) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$601.13 - $1,371.09
Chemical Identifiers
| CAS | 107552-14-7 |
|---|---|
| Molecular Formula | N3O9Sc |
| Molecular Weight (g/mol) | 230.97 |
| MDL Number | MFCD00011220 |
| InChI Key | DFCYEXJMCFQPPA-UHFFFAOYSA-N |
| PubChem CID | 60160869 |
| IUPAC Name | scandium(3+) trinitrate |
| SMILES | [Sc+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010048100
|
Ricca Chemical Company
RV010048100N |
100 mL |
N/A
|
|
|||||
|
RV010048500
|
Ricca Chemical Company
RV010048500N |
500 mL |
N/A
|
|
|||||
Description
- 1000 ppm in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 107552-14-7 | |
| 230.97 | |
| DFCYEXJMCFQPPA-UHFFFAOYSA-N | |
| scandium(3+) trinitrate |
| N3O9Sc | |
| MFCD00011220 | |
| 60160869 | |
| [Sc+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
| Natural LDPE Bottle | |
| 95.54%,1.96%,0.50% | |
| 1.01 | |
| N3O9Sc | |
| Approximately 0°C | |
| [Sc+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 230.97 | |
| 60160869 |
| 107552-14-7,7697-37-2,7732-18-5 | |
| 99.44%,2.04%,0.52% | |
| Liquid | |
| MFCD00011220 | |
| DFCYEXJMCFQPPA-UHFFFAOYSA-N | |
| scandium(3+) trinitrate | |
| 100 mL | |
| High Purity |