missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Samarium Standard for ICP 10 g/L in 2% HNO3, Ricca Chemical

Catalog No. RV010416100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010416100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RV010416100 Supplier Ricca Chemical Company Supplier No. RV010416100N

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 10 g/L in 2% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Specifications

Packaging Natural LDPE Bottle
CAS 13759-83-6, 7732-18-5
CAS Min % 93.84%, 2.20%, 1.96%
CAS Max % 97.68%, 2.28%, 2.04%
Specific Gravity 1
Physical Form Liquid
Molecular Formula H12N3O15Sm
Color Colorless
Melting Point Approximately 0°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Standard
InChI Key HDCOFJGRHQAIPE-UHFFFAOYSA-N
SMILES O.O.O.O.O.O.[Sm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
IUPAC Name samarium(3+) hexahydrate trinitrate
Molecular Weight (g/mol) 444.46
Quantity 100 mL
PubChem CID 203081
Grade High Purity
Chemical Name or Material Samarium ICP Standard
Show More Show Less