missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Rhodium (Rh) Standard for AAS 1000 ppm in 2% HCl, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$1013.09 - $4699.63
Chemical Identifiers
| CAS | 10049-07-7 |
|---|---|
| Molecular Formula | Cl3Rh |
| Molecular Weight (g/mol) | 209.26 |
| MDL Number | MFCD00011204 |
| InChI Key | SONJTKJMTWTJCT-UHFFFAOYSA-K |
| IUPAC Name | rhodium(3+) trichloride |
| SMILES | [Cl-].[Cl-].[Cl-].[Rh+3] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010044100
|
Ricca Chemical Company
RV010044100C |
100 mL |
Each for $1,013.09
|
|
|||||
|
RV010044500
|
Ricca Chemical Company
RV010044500C |
500 mL |
Each for $4,699.63
|
|
|||||
Description
- 1000 ppm in 2% HCl
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 10049-07-7 | |
| 209.26 | |
| SONJTKJMTWTJCT-UHFFFAOYSA-K | |
| [Cl-].[Cl-].[Cl-].[Rh+3] |
| Cl3Rh | |
| MFCD00011204 | |
| rhodium(3+) trichloride |
Specifications
| Amber Glass Bottle | |
| 92.90%,4.90%,0.20% | |
| 1.027 | |
| Cl3Rh | |
| UN3264 | |
| SONJTKJMTWTJCT-UHFFFAOYSA-K | |
| rhodium(3+) trichloride | |
| 100 mL |
| 10049-07-7,7647-01-0,7732-18-5 | |
| 96.70%,5.10%,0.20% | |
| Liquid | |
| MFCD00011204 | |
| Approximately 0°C | |
| [Cl-].[Cl-].[Cl-].[Rh+3] | |
| 209.26 | |
| High Purity |