missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Oxalate - C2O42- Standard for Ion Chromatography 1000 ppm in H2O, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Facility
$362.78 - $705.84
Chemical Identifiers
| CAS | 1113-38-8 |
|---|---|
| Molecular Formula | C2H8N2O4 |
| Molecular Weight (g/mol) | 124.096 |
| InChI Key | VBIXEXWLHSRNKB-UHFFFAOYSA-N |
| PubChem CID | 14213 |
| IUPAC Name | diazanium;oxalate |
| SMILES | C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010933100
|
Ricca Chemical Company
RV010933100N |
100 mL |
Each for $362.78
|
|
|||||
|
RV010933250
|
Ricca Chemical Company
RV010933250N |
250 mL |
Each for $493.97
|
|
|||||
|
RV010933500
|
Ricca Chemical Company
RV010933500N |
500 mL |
Each for $705.84
|
|
|||||
Description
- 1000 ppm in H2O
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 1113-38-8 | |
| 124.096 | |
| 14213 | |
| C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+] |
| C2H8N2O4 | |
| VBIXEXWLHSRNKB-UHFFFAOYSA-N | |
| diazanium;oxalate |
Specifications
| Natural LDPE Bottle | |
| 97.86%,0.14% | |
| 1.01 | |
| C2H8N2O4 | |
| VBIXEXWLHSRNKB-UHFFFAOYSA-N | |
| diazanium;oxalate | |
| 100 mL | |
| High Purity |
| 1113-38-8,7732-18-5 | |
| 100%,0.14% | |
| Liquid | |
| Approximately 0°C | |
| C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+] | |
| 124.096 | |
| 14213 |