missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Multi-Element Standard 9, Ricca Chemical

Catalog No. RV010715100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010715100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RV010715100 Supplier Ricca Chemical Company Supplier No. RV010715100N

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

100 ppm: As, Be, Pb, Cd, Cr, Ni, Hg, Se, Tl /nManufactured and Tested in an ISO 17025/Guide 34 Facil

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Specifications

Chemical Name or Material Multi Element Standard
Packaging Natural LDPE Bottle
CAS 7697-37-2, 13597-99-4, 13548-38-4, 13138-45-9, 7778-39-4, 7697-37-2
CAS Min % 4.90%, 0.15%, 0.05%, 0.03%, 0.02%
CAS Max % 5.10%, 0.15%, 0.05%, 0.03%, 0.02%
Specific Gravity 1.07
Physical Form Liquid
UN Number UN3264
Color Colorless
Melting Point Approximately 0°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Standard
Quantity 100 mL
Grade High Purity
Show More Show Less