missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Multi-Element Standard 4, Ricca Chemical

Catalog No. rv010706100
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010706100 100 mL
RV010706500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010706100 Supplier Ricca Chemical Company Supplier No. RV010706100N
Only null left

5000 ppm: Ca, K, Mg, Na /nManufactured and Tested in an ISO 17025/Guide 34 Facility

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Specifications

Packaging Natural LDPE Bottle
CAS Min % 4.90%, 2.99%, 2.01%, 1.81%, 1.26%
CAS Max % 5.10%, 3.11%, 2.09%, 1.89%, 1.32%
CAS 10377-60-3, 10124-37-5, 7631-99-4, 7757-79-1, 7697-37-2
Specific Gravity 1.06
Physical Form Liquid
UN Number UN3264
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Grade High Purity
Show More Show Less