missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Multi-Element Standard 4, Ricca Chemical
Description
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
| Chemical Name or Material | ICP Multi Element Standards |
| Packaging | Natural LDPE Bottle |
| CAS | 13473-90-0, 10377-60-3, 10124-37-5, 10421-48-4, 7697-37-2 |
| CAS Min % | 6.81%, 4.90%, 2.99%, 2.01%, 0.85% |
| CAS Max % | 7.09%, 5.10%, 3.11%, 2.09%, 0.89% |
| Specific Gravity | 1.06 |
| Physical Form | Liquid |
| UN Number | UN3264 |
| Color | Colorless |
| Melting Point | Approximately 0°C |
| Show More |