missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Multi-Element Standard 23, Ricca Chemical

Catalog No. RV010826100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010826100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RV010826100 Supplier Ricca Chemical Company Supplier No. RV010826100N

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

15K ppm Mg, 8K ppm Na, 3.5K ppm Ca, 3000 ppm K; 100 ppb B, Fe, Mo, Sr; 50 ppb As, Ba, Ni, V, Zn

  • 15,000ppm Mg, 8,000ppm Na, 3500ppm Ca, 3000ppm K, 100ppb B, Fe, Mo, Sr; 50ppb As, Ba, Ni, V, Zn; 30ppb Mn; 25ppb Co, Pb; 20ppb Be, Cd, Cr, Cu; 10ppb Se, Tl
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Specifications

Chemical Name or Material Multi Element Standard
Packaging Natural LDPE Bottle
CAS 7732-18-5
Specific Gravity 1.09
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type ICP Multi-element Standard Solution
Quantity 100 mL
Grade High Purity
Show More Show Less