missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Molybdenum Standard for ICP/MS 10 ppm in H2O, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Supplier: Ricca Chemical Company RV010603100N
Description
- 10 ppm in H2O
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
| Molybdenum ICP-MS Standard | |
| 12027-67-7,7732-18-5 | |
| 100%,0.00% | |
| Liquid | |
| MFCD00064633 | |
| Approximately 0°C | |
| Miscible with water | |
| Standard | |
| [NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].O=[Mo]123(=O)O[Mo]45(=O)(=O)O[Mo]67(=O)(=O)O[Mo]89(=O)(=O)O[Mo]%10%11(=O)(=O)O[Mo]%12(=O)(=O)(O1)[O+]2[Mo]([O+]34)([O+]56)([O+]%10%12)([O+]78)[O+]9%11 | |
| 1163.85 | |
| 71306766 |
| Natural LDPE Bottle | |
| 98%,0.00% | |
| 1 | |
| H24Mo7N6O24 | |
| Colorless | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Calibration Standard | |
| MISTUIVWPMGXFX-UHFFFAOYSA-T | |
| hexaammonium dodecaoxododecacyclo[7.5.1.13,5.13,13.15,7.17,9.01,4.01,6.01,8.02,13.011,14.011,15]heptamolybdoxane-2,4,6,8,14,15-hexakis(ylium) | |
| 100 mL | |
| High Purity |