missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Mixed Anion Standard 7, Ricca Chemical

150 ppm SO42-, PO43-; 100 ppm NO3-, Br-, NO2-; 30 ppm Cl-, 20 ppm F- /nManufactured and

$681.91 - $1193.09

Specifications

CAS Min % 0.98%,0.12%,0.02%,0.01%,0.00%
CAS Max % 1.02%,0.12%,0.02%,0.01%,0.00%
CAS 7722-76-1,7783-20-2,7631-99-4,7647-14-5,7732-18-5
Specific Gravity 1.07
Physical Form Liquid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RV01068550N
View Documents
Ricca Chemical Company
RV01068550N
50 mL
Each for $681.91
Only null left
 
RV010685100
View Documents
Ricca Chemical Company
RV010685100N
100 mL
Each for $1,193.09
Only null left
 
Description

Description

  • 150 ppm SO42-, PO43; 100 ppm NO3-, Br-, NO2-; 30 ppm Cl-, 20 ppm F-
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

0.98%,0.12%,0.02%,0.01%,0.00%
7722-76-1,7783-20-2,7631-99-4,7647-14-5,7732-18-5
Liquid
High Purity
1.02%,0.12%,0.02%,0.01%,0.00%
1.07
Approximately 0°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'