missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Lithium Standard for ICP/MS 10 ppm in 2% HNO3, Ricca Chemical

Catalog No. RV010599100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010599100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RV010599100 Supplier Ricca Chemical Company Supplier No. RV010599100N

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 10 ppm in 2% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Specifications

Packaging Natural LDPE Bottle
CAS Min % 96.03%, 1.96%, 0.01%
CAS Max % 99.95%, 2.04%, 0.01%
CAS 7697-37-2, 7732-18-5
Specific Gravity 1
Physical Form Liquid
Molecular Formula LiNO3
MDL Number MFCD00011094
Color Colorless
Melting Point Approximately 0°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Standard
InChI Key IIPYXGDZVMZOAP-UHFFFAOYSA-N
SMILES [Li+].[O-][N+]([O-])=O
IUPAC Name lithium(1+) nitrate
Molecular Weight (g/mol) 68.94
Quantity 100 mL
PubChem CID 10129889
ChEBI CHEBI:63315
Grade High Purity
Chemical Name or Material Lithium ICP-MS Standard
Show More Show Less