missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Lithium Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Supplier:  Ricca Chemical Company RV010341250N

 View more versions of this product

Catalog No. RV010341250


Only null left
Explore available promotions

Description

Description

  • 1000 ppm in 2% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

Natural Poly Bottle
95.07%,1.96%,0.97%
1
LiNO3
Approximately 0°C
IIPYXGDZVMZOAP-UHFFFAOYSA-N
lithium(1+) nitrate
250 mL
CHEBI:63315
7790-69-4,7697-37-2,7732-18-5
98.95%,2.04%,1.01%
Liquid
MFCD00011094
Standard
[Li+].[O-][N+]([O-])=O
68.94
10129889
High Purity

Chemical Identifiers

7790-69-4
68.94
IIPYXGDZVMZOAP-UHFFFAOYSA-N
CHEBI:63315
[Li+].[O-][N+]([O-])=O
LiNO3
MFCD00011094
10129889
lithium(1+) nitrate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions