missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Lanthanum Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Supplier: Ricca Chemical Company RV010289250N
Description
- 1000 ppm in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
| Natural Poly Bottle | |
| 95.74%,1.96%,0.30% | |
| 1 | |
| H12LaN3O15 | |
| Approximately 0°C | |
| GJKFIJKSBFYMQK-UHFFFAOYSA-N | |
| lanthanum(3+) hexahydrate trinitrate | |
| 250 mL | |
| High Purity |
| 10277-43-7,7697-37-2,7732-18-5 | |
| 99.64%,2.04%,0.32% | |
| Liquid | |
| MFCD00149751 | |
| Standard | |
| O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 433.01 | |
| 16211478 |
Chemical Identifiers
| 10277-43-7 | |
| 433.01 | |
| GJKFIJKSBFYMQK-UHFFFAOYSA-N | |
| lanthanum(3+) hexahydrate trinitrate |
| H12LaN3O15 | |
| MFCD00149751 | |
| 16211478 | |
| O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |