missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Iridium Standard for ICP/MS 10 ppm in 2% HCl, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Supplier: Ricca Chemical Company RV010596100N
Description
- 10 ppm in 2% HCl
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
| Iridium ICP-MS Standard | |
| 10025-83-9,7647-01-0,7732-18-5 | |
| 99.96%,2.04%,0.00% | |
| Liquid | |
| Colorless | |
| Approximately 0°C | |
| Miscible with water | |
| Standard | |
| Cl[Ir](Cl)Cl | |
| 298.567 | |
| 25563 |
| Natural LDPE Bottle | |
| 96.04%,1.96%,0.00% | |
| 1 | |
| Cl3Ir | |
| <2 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Calibration Standard | |
| DANYXEHCMQHDNX-UHFFFAOYSA-K | |
| trichloroiridium | |
| 100 mL | |
| High Purity |