missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Iridium Standard for ICP 10,000 ppm in 10% HCl, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Supplier:  Ricca Chemical Company RV010389100N

Catalog No. RV010389100


Only null left
Explore available promotions

Description

Description

  • 10,000 ppm in 10% HCl
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

Iridium ICP Standard
10025-83-9,7647-01-0,7732-18-5
90.22%,10.20%,1.58%
Liquid
UN3264
<2
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Calibration Standard
DANYXEHCMQHDNX-UHFFFAOYSA-K
trichloroiridium
100 mL
High Purity
Natural LDPE Bottle
86.68%,9.80%,1.52%
1
Cl3Ir
Colorless
Approximately 0°C
Miscible with water
Standard
Cl[Ir](Cl)Cl
298.567
25563
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions