missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Internal Calibration Standard 7, Ricca Chemical

Catalog No. RV010858100
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010858100 100 mL
RV010858500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010858100 Supplier Ricca Chemical Company Supplier No. RV010858100N

10 ppm: 6Li, Sc, Y, In, Tb, Bi /nManufactured and Tested in an ISO 17025/Guide 34 Facility

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Specifications

Packaging Natural LDPE Bottle
CAS Min % 4.90%, 0.01%
CAS Max % 5.10%, 0.01%
CAS 7790-69-4, 10035-06-0, 7697-37-2
Specific Gravity 1.06
Physical Form Liquid
UN Number UN3264
Melting Point Approximately 0°C
Quantity 100 mL
Grade High Purity