missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Interference Check Solution 12, Ricca Chemical

Catalog No. RV010796500
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010796500 500 mL
RV010796100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010796500 Supplier Ricca Chemical Company Supplier No. RV010796500N

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

10K ppm Cl, 2000 ppm C; 1000 ppm Al, Ca, Fe, K, Mg, Na, P, S; 20 ppm Mo, Ti /nManufactured and Teste

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Specifications

Packaging Natural Poly Bottle
CAS 7697-37-2, 13473-90-0
CAS Min % 4.90%, 2.72%
CAS Max % 5.10%, 2.84%
Specific Gravity 1.07
Physical Form Liquid
UN Number UN3264
Melting Point Approximately 0°C
Quantity 500 mL
Grade High Purity