missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ ICP Tuning Solution 15, Ricca Chemical

Catalog No. RV010727500
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010727500 500 mL
RV010727100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010727500 Supplier Ricca Chemical Company Supplier No. RV010727500N

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

500 ppm: K; 50 ppm: Al, As, Ba, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Se, Sr, Zn /nManufactured and Tested

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Specifications

Packaging Natural Poly Bottle
CAS 7697-37-2, 13473-90-0, 13548-38-4, 3251-23-8
CAS Min % 0.98%, 0.07%, 0.02%, 0.01%
CAS Max % 1.02%, 0.07%, 0.02%, 0.01%
Specific Gravity 1.08
Physical Form Liquid
Melting Point Approximately 0°C
Quantity 500 mL
Grade High Purity