missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Holmium Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$572.60 - $1479.03
Chemical Identifiers
| CAS | 14483-18-2 |
|---|---|
| Molecular Formula | H10HoN3O14 |
| Molecular Weight (g/mol) | 441.017 |
| InChI Key | BJUGWWDCFYEYOA-UHFFFAOYSA-N |
| PubChem CID | 25021754 |
| IUPAC Name | holmium(3+);trinitrate;pentahydrate |
| SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.[Ho+3] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010285100
|
Ricca Chemical Company
RV010285100N |
100 mL |
Each for $572.60
|
|
|||||
|
RV010285250
|
Ricca Chemical Company
RV010285250N |
250 mL |
Each for $905.80
|
|
|||||
|
RV010285500
|
Ricca Chemical Company
RV010285500N |
500 mL |
Each for $1,479.03
|
|
|||||
Description
- 1000 ppm in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 14483-18-2 | |
| 441.017 | |
| 25021754 | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.[Ho+3] |
| H10HoN3O14 | |
| BJUGWWDCFYEYOA-UHFFFAOYSA-N | |
| holmium(3+);trinitrate;pentahydrate |
Specifications
| Natural LDPE Bottle | |
| 95.78%,1.96%,0.26% | |
| 1 | |
| H10HoN3O14 | |
| Standard | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.[Ho+3] | |
| 441.017 | |
| 25021754 |
| 14483-18-2,7697-37-2,7732-18-5 | |
| 99.68%,2.04%,0.28% | |
| Liquid | |
| Approximately 0°C | |
| BJUGWWDCFYEYOA-UHFFFAOYSA-N | |
| holmium(3+);trinitrate;pentahydrate | |
| 100 mL | |
| High Purity |