missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Elan DRC Plus Tuning Solution II, Ricca Chemical

10 ppb: Ba; 1 ppb: Be, Ce, Co, In, Fe, Pb, Mg, Th, U /nManufactured and Tested in an ISO 17025/Guide

$981.13 - $1610.41

Specifications

CAS 7697-37-2,13823-29-5
CAS Min % 0.49%
CAS Max % 0.51%
Specific Gravity 1.07
Physical Form Liquid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RV010878100
View Documents
Ricca Chemical Company
RV010878100N
100 mL
Each for $1,610.41
Only null left
 
RV010878500
View Documents
Ricca Chemical Company
RV010878500N
500 mL
Each for $981.13
Only null left
 
Description

Description

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

7697-37-2,13823-29-5
0.51%
Liquid
High Purity
0.49%
1.07
Approximately 0°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'