missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Elan DRC Plus Tuning Solution II, Ricca Chemical

Catalog No. RV010878100
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010878100 100 mL
RV010878500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010878100 Supplier Ricca Chemical Company Supplier No. RV010878100N

10 ppb: Ba; 1 ppb: Be, Ce, Co, In, Fe, Pb, Mg, Th, U /nManufactured and Tested in an ISO 17025/Guide

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Specifications

Packaging Natural LDPE Bottle
CAS 7697-37-2, 13823-29-5
CAS Min % 0.49%
CAS Max % 0.51%
Specific Gravity 1.07
Physical Form Liquid
Melting Point Approximately 0°C
Quantity 100 mL
Grade High Purity