missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Aluminum (Al) Standard for AAS 1000 ppm in 2% HCl, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Supplier: Ricca Chemical Company RV010002500N
Includes: Detailed Certificate of Analysis
Specifications
| Aluminum (Al) Standard for AAS | |
| 7784-27-2,7647-01-0,7446-70-0,7732-18-5 | |
| 0.994602,0.0204,0.004998 | |
| Liquid | |
| Approximately 0°C | |
| SWCIQHXIXUMHKA-UHFFFAOYSA-N | |
| aluminum;trinitrate;nonahydrate | |
| 375.129 | |
| 24567 |
| Natural Poly Bottle | |
| 0.955598,0.0196,0.004802 | |
| 1.013 | |
| AlH18N3O18 | |
| Standard | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] | |
| Atomic Absorption | |
| 500 mL | |
| High Purity |
Chemical Identifiers
| 7784-27-2 | |
| 375.129 | |
| 24567 | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] |
| AlH18N3O18 | |
| SWCIQHXIXUMHKA-UHFFFAOYSA-N | |
| aluminum;trinitrate;nonahydrate |