missing translation for 'onlineSavingsMsg'
Learn More

Veratryl Alcohol 98.0+%, TCI America™

Catalog No. v002025ml
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V002025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V002025ML Supplier TCI America Supplier No. V002025ML

Chemical Identifiers

CAS 93-03-8
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.192
MDL Number MFCD00004638
InChI Key OEGPRYNGFWGMMV-UHFFFAOYSA-N
Synonym 3,4-dimethoxybenzyl alcohol, veratryl alcohol, 3,4-dimethoxyphenyl methanol, benzenemethanol, 3,4-dimethoxy, 3,4-dimethoxyphenylmethyl alcohol, veratrole alcohol, 3,4-dimethoxybenzenemethanol, unii-mb4t4a711h, 3,4-dimethoxyphenyl-methanol
PubChem CID 7118
ChEBI CHEBI:62150
IUPAC Name (3,4-dimethoxyphenyl)methanol
SMILES COC1=C(C=C(C=C1)CO)OC

Specifications

Color Yellow
Boiling Point 173°C
Quantity 25 mL
Formula Weight 168.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Veratryl Alcohol
EINECSNumber (3)-1078
TSCA Yes