missing translation for 'onlineSavingsMsg'
Learn More

Veratrole, 99%

Catalog No. p-7081830
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1198530 250 g
AAA119850B 1000 g
AAA119850I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1198530 Supplier Thermo Scientific Chemicals Supplier No. A1198530

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 91-16-7 | C8H10O2 | 138.166 g/mol

It is useful for organic synthesis of aromatic compounds. Veratrole is relatively electron-rich and thus readily undergoes electrophilic substitution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is useful for organic synthesis of aromatic compounds. Veratrole is relatively electron-rich and thus readily undergoes electrophilic substitution.

Solubility
Soluble in alcohol, diethyl ether, acetone, and methanol. Slightly soluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.

Chemical Identifiers

CAS 91-16-7
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00008357
InChI Key ABDKAPXRBAPSQN-UHFFFAOYSA-N
Synonym veratrole, veratrol, pyrocatechol dimethyl ether, o-dimethoxybenzene, catechol dimethyl ether, benzene, 1,2-dimethoxy, 2-methoxyanisole, o,o-dimethyl catechol, 2-dimethoxybenzol, benzene, o-dimethoxy
PubChem CID 7043
ChEBI CHEBI:59114
IUPAC Name 1,2-dimethoxybenzene
SMILES COC1=CC=CC=C1OC

Specifications

Melting Point 21°C to 23°C
Density 1.084
Boiling Point 206°C to 207°C
Flash Point 87°C (188°F)
Refractive Index 1.533
Quantity 250 g
Beilstein 1364621
Merck Index 14,9956
Solubility Information Soluble in alcohol,diethyl ether,acetone,and methanol. Slightly soluble in water.
Formula Weight 138.17
Percent Purity 99%
Chemical Name or Material Veratrole
Show More Show Less
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 202-045-3
RTECSNumber CZ6475000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only