missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Verapamil Hydrochloride 98.0+%, TCI America™
Supplier: TCI America V01185G
Specifications
| Verapamil Hydrochloride | |
| 145°C | |
| 5.25 (0.1% aq. soln.) | |
| MFCD00055208 | |
| 2811 | |
| DOQPXTMNIUCOSY-UHFFFAOYSA-N | |
| 2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-propan-2-ylpentanenitrile;hydrochloride | |
| 62969 | |
| ≥98.0% (HPLC,T) |
| 152-11-4 | |
| White | |
| C27H39ClN2O4 | |
| 5 g | |
| verapamil hydrochloride, verapamil hcl, manidon, calcan hydrochloride, cardibeltin, cardiabeltin, cardioprotect, veroptinstada, calaptin, caveril | |
| CC(C)C(CCCN(C)CCC1=CC(=C(C=C1)OC)OC)(C#N)C2=CC(=C(C=C2)OC)OC.Cl | |
| 491.069 | |
| 491.07 | |
| Crystalline Powder |
Chemical Identifiers
| 152-11-4 | |
| 491.069 | |
| DOQPXTMNIUCOSY-UHFFFAOYSA-N | |
| 62969 | |
| CC(C)C(CCCN(C)CCC1=CC(=C(C=C1)OC)OC)(C#N)C2=CC(=C(C=C2)OC)OC.Cl |
| C27H39ClN2O4 | |
| MFCD00055208 | |
| verapamil hydrochloride, verapamil hcl, manidon, calcan hydrochloride, cardibeltin, cardiabeltin, cardioprotect, veroptinstada, calaptin, caveril | |
| 2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-propan-2-ylpentanenitrile;hydrochloride |
Safety and Handling
RTECSNumber : YV8320000
TSCA : No