Learn More
Thermo Scientific Chemicals (±)-Verapamil hydrochloride, 99+%
CAS: 152-11-4 | C27H38N2O4·HCl | 491.07 g/mol
$171.51 - $171.51
Chemical Identifiers
| CAS | 152-11-4 |
|---|---|
| Molecular Formula | C27H38N2O4·HCl |
| Molecular Weight (g/mol) | 491.07 |
| MDL Number | MFCD00055208 |
| InChI Key | DOQPXTMNIUCOSY-UHFFFAOYSA-N |
| Synonym | verapamil hydrochloride, verapamil hcl, manidon, calcan hydrochloride, cardibeltin, cardiabeltin, cardioprotect, veroptinstada, calaptin, caveril |
| PubChem CID | 62969 |
| IUPAC Name | 2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-propan-2-ylpentanenitrile;hydrochloride |
| SMILES | CC(C)C(CCCN(C)CCC1=CC(=C(C=C1)OC)OC)(C#N)C2=CC(=C(C=C2)OC)OC.Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC329330010
|
Thermo Scientific Chemicals
329330010 |
1 g | Glass bottle |
Each for $171.51
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 152-11-4 | |
| 491.07 | |
| DOQPXTMNIUCOSY-UHFFFAOYSA-N | |
| 62969 | |
| CC(C)C(CCCN(C)CCC1=CC(=C(C=C1)OC)OC)(C#N)C2=CC(=C(C=C2)OC)OC.Cl |
| C27H38N2O4·HCl | |
| MFCD00055208 | |
| verapamil hydrochloride, verapamil hcl, manidon, calcan hydrochloride, cardibeltin, cardiabeltin, cardioprotect, veroptinstada, calaptin, caveril | |
| 2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-propan-2-ylpentanenitrile;hydrochloride |
Specifications
| 152-11-4 | |
| 100.0 | |
| White | |
| 0.1% max. | |
| Authentic | |
| Glass bottle | |
| MFCD00055208 | |
| 15, 10144 | |
| Solubility in water: soluble in water. Other solubilities: soluble in ethanol | |
| CC(C)C(CCCN(C)CCC1=CC(=C(C=C1)OC)OC)(C#N)C2=CC(=C(C=C2)OC)OC.Cl | |
| 491.07 | |
| 491.07 | |
| Powder |
| 99.0 | |
| 140.0°C to 144.0°C | |
| 4.5 to 6.5 (5% soln.) | |
| 0.5% max. (105°C, 2 hrs) | |
| 99+% | |
| C27H38N2O4·HCl | |
| 1 g | |
| verapamil hydrochloride, verapamil hcl, manidon, calcan hydrochloride, cardibeltin, cardiabeltin, cardioprotect, veroptinstada, calaptin, caveril | |
| DOQPXTMNIUCOSY-UHFFFAOYSA-N | |
| 2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-propan-2-ylpentanenitrile;hydrochloride | |
| 62969 | |
| 99+% | |
| (±)-Verapamil hydrochloride |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
Toxic in contact with skin.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Danger
EINECSNumber : 205-800-5
RUO – Research Use Only