missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (±)-Verapamil hydrochloride, 99+%

Catalog No. AC329330010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC329330010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC329330010 Supplier Thermo Scientific Chemicals Supplier No. 329330010
Only null left

CAS: 152-11-4 | C27H38N2O4·HCl | 491.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 152-11-4
Molecular Formula C27H38N2O4·HCl
Molecular Weight (g/mol) 491.07
MDL Number MFCD00055208
InChI Key DOQPXTMNIUCOSY-UHFFFAOYSA-N
Synonym verapamil hydrochloride, verapamil hcl, manidon, calcan hydrochloride, cardibeltin, cardiabeltin, cardioprotect, veroptinstada, calaptin, caveril
PubChem CID 62969
IUPAC Name 2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-propan-2-ylpentanenitrile;hydrochloride
SMILES CC(C)C(CCCN(C)CCC1=CC(=C(C=C1)OC)OC)(C#N)C2=CC(=C(C=C2)OC)OC.Cl

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 140.0°C to 144.0°C
Color White
pH 4.5 to 6.5 (5% soln.)
Ignition Residue 0.1% max.
Loss on Drying 0.5% max. (105°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 10144
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in ethanol
Formula Weight 491.07
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material (±)-Verapamil hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
Toxic in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
EINECSNumber 205-800-5

RUO – Research Use Only