Learn More
MSA™ Ventilation Smoke Tube Kits
Catalog No. 189993391
Change view
Click to view available options
Type:
Smoke Tube
Smoke Tube Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
189993391 Smoke Tube
189993414 Smoke Tube Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 189993391 Supplier MSA™ Supplier No. 458480
  • Allows for controlled generation of visible smoke
  • Determines velocity and flow patterns of heating, ventilating and air-conditioning systems
  • Available with glass or plastic tubes with break-off tips
Use these kits to determine the velocity of slow-moving air currents and establish their direction and flow patterns. MSA™ Ventilation Smoke Tube Kits are ideal for use in commercial buildings and industrial processing plants as well as in shafts, mines and tunnels.
  • Allows for controlled generation of visible smoke
  • Determines velocity and flow patterns of heating, ventilating and air-conditioning systems
  • Available with glass or plastic tubes with break-off tips
Specifications
Description Box of twelve ventilation smoke tubes
For Use With (Equipment) Ventilation Smoke Tube Kits
Type Smoke Tube
Material Plastic
Quantity 12
Tip Type Break-off