missing translation for 'onlineSavingsMsg'
Learn More

Venlafaxine Hydrochloride 98.0+%, TCI America™

Catalog No. V01101G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V01101G 1 g
V01105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V01101G Supplier TCI America Supplier No. V01101G

Chemical Identifiers

CAS 99300-78-4
Molecular Formula C17H28ClNO2
Molecular Weight (g/mol) 313.866
MDL Number MFCD03658865
InChI Key QYRYFNHXARDNFZ-UHFFFAOYSA-N
Synonym venlafaxine hydrochloride, effexor, venlafaxine hcl, effexor xr, trevilor, dobupal, efexor, vandral, venlafaxinehydrochloride
PubChem CID 62923
ChEBI CHEBI:9944
IUPAC Name 1-[2-(dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexan-1-ol;hydrochloride
SMILES CN(C)CC(C1=CC=C(C=C1)OC)C2(CCCCC2)O.Cl

Specifications

Melting Point 217°C
Color White
Quantity 1 g
Formula Weight 313.87
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Venlafaxine Hydrochloride
RTECSNumber QV8872760
TSCA No