missing translation for 'onlineSavingsMsg'
Learn More

Venlafaxine Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111016016
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016016 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016016 Supplier Merck Emd Millipore Supplier No. PHR17361G
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Venlafaxine Hydrochloride is the hydrochloride salt of venlafaxine, a non-tricyclic antidepressant and serotonin-norepinephrine-dopamine reuptake inhibitor.

Specifications

CAS 99300-78-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C17H27 NO2 · HCl
MDL Number MFCD03658865
Synonym Venlafaxine hydrochloride; (+/-)-1-[2-(Dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride; Effexor
Type Certified Reference Material
Quantity 1 g
Formula Weight 313.86
Grade Certified Reference Material