missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mopec Vein Thrombus Control Slides
Normal Human Tissue Vein Thrombus Control Slides
$2,278.21 - $4,591.25
Specifications
| Type | Control Slide |
|---|---|
| For Use With (Application) | Use as a Control |
| Content And Storage | Room-Temperature |
Description
- High-quality control slides to support accurate histopathology and laboratory testing
- Includes well-preserved normal human vein tissue for reliable comparison and analysis
- Easy-to-use slides suitable for both laboratory and educational purposes
- Prepared to maintain tissue integrity for consistent experimental and diagnostic results
Specifications
| Control Slide | |
| Use as a Control | |
| Room-Temperature |