missing translation for 'onlineSavingsMsg'
Learn More

Mopec Vein Thrombus Control Slides

Catalog No. 21010276
Change view
Click to view available options
Quantity:
100 Control Slides
25 Control Slides
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21010276 25 Control Slides
21010275 100 Control Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21010276 Supplier Mopec Supplier No. SC95825
Only null left

Normal Human Tissue Vein Thrombus Control Slides

  • High-quality control slides to support accurate histopathology and laboratory testing
  • Includes well-preserved normal human vein tissue for reliable comparison and analysis
  • Easy-to-use slides suitable for both laboratory and educational purposes
  • Prepared to maintain tissue integrity for consistent experimental and diagnostic results

Specifications

Content And Storage Room-Temperature
Quantity 25 Control Slides
Type Control Slide
For Use With (Application) Use as a Control
Includes 24 Unstained 1 Stained Control Slide