missing translation for 'onlineSavingsMsg'
Learn More
Learn More
vBRIDP(regR) 98.0+%, TCI America™
$74.56 - $223.68
Chemical Identifiers
| CAS | 384842-25-5 |
|---|---|
| Molecular Formula | C23H31P |
| Molecular Weight (g/mol) | 338.475 |
| MDL Number | MFCD16621412 |
| InChI Key | CYGZMOQFIMJEIT-UHFFFAOYSA-N |
| Synonym | Di-tert-butyl(1,1-diphenyl-1-propen-2-yl)phosphine |
| PubChem CID | 12010825 |
| IUPAC Name | ditert-butyl(1,1-diphenylprop-1-en-2-yl)phosphane |
| SMILES | CC(=C(C1=CC=CC=C1)C2=CC=CC=C2)P(C(C)(C)C)C(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
V0132200MG
|
TCI America
V0132200MG |
200 mg |
Each for $74.56
|
|
|||||
|
V01321G
|
TCI America
V01321G |
1 g |
Each for $223.68
|
|
|||||
Chemical Identifiers
| 384842-25-5 | |
| 338.475 | |
| CYGZMOQFIMJEIT-UHFFFAOYSA-N | |
| 12010825 | |
| CC(=C(C1=CC=CC=C1)C2=CC=CC=C2)P(C(C)(C)C)C(C)(C)C |
| C23H31P | |
| MFCD16621412 | |
| Di-tert-butyl(1,1-diphenyl-1-propen-2-yl)phosphine | |
| ditert-butyl(1,1-diphenylprop-1-en-2-yl)phosphane |
Specifications
| 384842-25-5 | |
| White-Yellow | |
| MFCD16621412 | |
| Di-tert-butyl(1,1-diphenyl-1-propen-2-yl)phosphine | |
| CC(=C(C1=CC=CC=C1)C2=CC=CC=C2)P(C(C)(C)C)C(C)(C)C | |
| 338.475 | |
| 338.47 | |
| Crystalline Powder |
| 133°C | |
| C23H31P | |
| 200 mg | |
| CYGZMOQFIMJEIT-UHFFFAOYSA-N | |
| ditert-butyl(1,1-diphenylprop-1-en-2-yl)phosphane | |
| 12010825 | |
| ≥98.0% (HPLC) | |
| vBRIDP(regR) |
Safety and Handling
TSCA : No