Learn More
Vasopressin, 98+%
CAS: 11000-17-2 | C46H65N15O12S2
$813.81 - $2494.37
Chemical Identifiers
| CAS | 11000-17-2 |
|---|---|
| Molecular Formula | C46H65N15O12S2 |
| MDL Number | MFCD00076738 |
| Synonym | Cys-Tyr-Phe-Gln-Asn-Cys-Pro-Arg-Gly-NH2; AVP peptide |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ61248MB
|
Thermo Scientific Chemicals
J61248MB |
25 mg |
Each for $813.81
|
|
|||||
|
AAJ61248MC
|
Thermo Scientific Chemicals
J61248MC |
100 mg |
Each for $2,494.37
|
|
|||||
Description
It is used as a peptide hormone that regulates the bodyâs retention of water. Vasopressin or antidiuretic hormone is a potent endogenous hormone which is responsible for regulating plasma osmolality and volume. It acts as a neurotransmitter in the brain to control circadian rhythm, thermoregulation, and adrenocorticotrophic hormone release (ACTH). The therapeutic use of vasopressin has become increasingly important in the critical care environment in the management of cranial diabetes insipidus, bleeding abnormalities, oesophageal variceal haemorrhage, asystolic cardiac arrest, and septic shock.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water (Miscible).
Notes
Store in airtight containers at 2° to 8°. Store in cool, sealed container.
Chemical Identifiers
| 11000-17-2 | |
| C46H65N15O12S2 | |
| MFCD00076738 | |
| Cys-Tyr-Phe-Gln-Asn-Cys-Pro-Arg-Gly-NH2; AVP peptide |
Specifications
| 11000-17-2 | |
| C46H65N15O12S2 | |
| 25 mg | |
| Soluble in water (Miscible). | |
| ≥98% | |
| Vasopressin |
| White | |
| MFCD00076738 | |
| Cys-Tyr-Phe-Gln-Asn-Cys-Pro-Arg-Gly-NH2; AVP peptide | |
| 1084.24 | |
| Lyophilized Powder |
Safety and Handling
EINECSNumber : 234-236-2
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only