accessibility menu, dialog, popup

UserName

CAS RN 11000-17-2

H2NHNONOSSH2NONHHOONHONHH2NOOHNNH2OOHNONHNH2OH2NSSHNONH2ONHOH2NONHONHOOHHNOONOHNNH2NH2NONHNH2O

CAS RN 11000-17-2

IUPAC Name: 2-({1-[19-amino-13-benzyl-10-(2-carbamoylethyl)-7-(carbamoylmethyl)-16-[(4-hydroxyphenyl)methyl]-6,9,12,15,18-pentaoxo-1,2-dithia-5,8,11,14,17-pentaazacycloicosane-4-carbonyl]pyrrolidin-2-yl}formamido)-N-(carbamoylmethyl)-5-[(diaminomethylidene)amino]pentanamide; 6-amino-2-({1-[19-amino-13-benzyl-10-(2-carbamoylethyl)-7-(carbamoylmethyl)-16-[(4-hydroxyphenyl)methyl]-6,9,12,15,18-pentaoxo-1,2-dithia-5,8,11,14,17-pentaazacycloicosane-4-carbonyl]pyrrolidin-2-yl}formamido)-N-(carbamoylmethyl)hexanamide
Synonyms: 2-({1-[19-amino-13-benzyl-10-(2-carbamoylethyl)-7-(carbamoylmethyl)-16-[(4-hydroxyphenyl)methyl]-6,9,12,15,18-pentaoxo-1,2-dithia-5,8,11,14,17-pentaazacycloicosane-4-carbonyl]pyrrolidin-2-yl}formamido)-N-(carbamoylmethyl)-5-[(diaminomethylidene)amino]pentanamide; 6-amino-2-({1-[19-amino-13-benzyl-10-(2-carbamoylethyl)-7-(carbamoylmethyl)-16-[(4-hydroxyphenyl)methyl]-6,9,12,15,18-pentaoxo-1,2-dithia-5,8,11,14,17-pentaazacycloicosane-4-carbonyl]pyrrolidin-2-yl}formamido)-N-(carbamoylmethyl)hexanamide
Molecular Weight (g/mol): 2140.46
Molecular Formula: C92H130N28O24S4
InChi Key: JLTCWSBVQSZVLT-UHFFFAOYNA-N
SMILES: NCCCCC(NC(=O)C1CCCN1C(=O)C1CSSCC(N)C(=O)NC(CC2=CC=C(O)C=C2)C(=O)NC(CC2=CC=CC=C2)C(=O)NC(CCC(N)=O)C(=O)NC(CC(N)=O)C(=O)N1)C(=O)NCC(N)=O.NC1CSSCC(NC(=O)C(CC(N)=O)NC(=O)C(CCC(N)=O)NC(=O)C(CC2=CC=CC=C2)NC(=O)C(CC2=CC=C(O)C=C2)NC1=O)C(=O)N1CCCC1C(=O)NC(CCCN=C(N)N)C(=O)NCC(N)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)