missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VASN Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5110822

Catalog No. PIPA5110822


Only null left

Description

Description

Immunogen sequence: LDVSNLSLQA LPGDLSGLFP RLRLLAAARN PFNCVCPLSW FGPWVRESHV TLASPEETRC HFPPKNAGRL LLELDYADFG CPATTTTATV PTTRPVVREP TALSSSLAPT Highest antigen sequence indentity to the following orthologs: Mouse - 86%, Rat - 90%.

The VASN gene encodes the protein vasorin, a type I transmembrane protein involved in cellular signaling, vascular development, and tissue repair. Vasorin is characterized by its expression in various tissues, including vascular endothelial cells and smooth muscle cells, where it functions to modulate cell proliferation and differentiation. It plays a significant role in the regulation of vascular remodeling and homeostasis by interacting with signaling molecules like transforming growth factor-beta (TGF-β), thereby influencing processes such as angiogenesis and wound healing. Vasorin is implicated in inhibiting TGF-β signaling pathways, which are crucial for maintaining vascular integrity and preventing excessive fibrosis during tissue repair. Dysregulation of VASN expression can contribute to pathological conditions, including cardiovascular diseases and cancer, where abnormal vascular remodeling and cell proliferation are evident. Research into vasorin continues to explore its functional mechanisms and potential therapeutic applications in diseases characterized by vascular dysregulation and tissue remodeling.
Specifications

Chemical Identifiers

VASN
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Human
VASN
2610528G05Rik; Atia; Protein slit-like 2; Slitl2; Slit-like 2; UNQ314/PRO357/PRO1282; VASN; Vasorin
VASN
Primary
Antigen affinity chromatography
0.05 mg/mL
Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q6EMK4
114990
Recombinant Protein corresonding to Human VASN. Recombinant protein control fragment (Product #RP-108312).
Antibody

Specifications

VASN
Polyclonal
Unconjugated
VASN
2610528G05Rik; Atia; Protein slit-like 2; Slitl2; Slit-like 2; UNQ314/PRO357/PRO1282; VASN; Vasorin
Rabbit
Antigen affinity chromatography
RUO
114990
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q6EMK4
VASN
Recombinant Protein corresonding to Human VASN. Recombinant protein control fragment (Product #RP-108312).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions