missing translation for 'onlineSavingsMsg'
Learn More

Variamine Blue B Base 98.0+%, TCI America™

Catalog No. M04931G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M04931G 1 g
M049325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M04931G Supplier TCI America Supplier No. M04931G
Only null left

Chemical Identifiers

CAS 101-64-4
Molecular Formula C13H14N2O
Molecular Weight (g/mol) 214.27
MDL Number MFCD00070553
InChI Key RBLUJIWKMSZIMK-UHFFFAOYSA-N
Synonym 4-Amino-4′C-methoxydiphenylamine, N-(4-Methoxyphenyl)-1,4-phenylenediamine
PubChem CID 66869
IUPAC Name N1-(4-methoxyphenyl)benzene-1,4-diamine
SMILES COC1=CC=C(NC2=CC=C(N)C=C2)C=C1

Specifications

Melting Point 101°C
Boiling Point 210°C
Formula Weight 214.27
Percent Purity ≥98.0% (T)
Quantity 1 g
Physical Form Crystal/Powder
Chemical Name or Material Variamine Blue B Base [Redox Indicator]
EINECSNumber (3)-2801
TSCA Yes